Tisk…
Gentamicin sulphate, Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
M-Clarity™ quality level = MQ200
G1264-1G G1264-250MG G1264-50MG G1264-5G
SIALG1264-1GEA 11090 CZK
SIALG1264-1G SIALG1264-250MG SIALG1264-50MG SIALG1264-5G
Gentamicin sulphate, Sigma-Aldrich®
Gentamicin sulphate
Bod tání: 218…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS číslo: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403