Tisk…

Gentamicin sulphate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

48760-1G-F 48760-25G-F 48760-5G-F
SIAL48760-1G-FEA 4740 CZK
SIAL48760-1G-F SIAL48760-25G-F SIAL48760-5G-F
Gentamicin sulphate, Sigma-Aldrich®
Gentamicin sulphate
Bod tání: 218…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS číslo: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR