L(-)-Mannose, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
M-Clarity™ quality level = MQ100
M1134-1G
M1134-500MG
SIALM1134-1GEA
18450
CZK
SIALM1134-1G
SIALM1134-500MG
L(-)-Mannose, Sigma-Aldrich®
L(-)-Mannose
Vzorec:
C₆H₁₂O₆ MW: 180,16 g/mol Bod varu: 376 °C (1013 hPa) Bod tání: 132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00136021 CAS číslo: 10030-80-5 EINECS: 233-080-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...