Tisk…

L(-)-Glucose, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

G5500-5G G5500-1G G5500-250MG G5500-25G G5500-500MG
SIALG5500-5GEA 16070 CZK
SIALG5500-5G SIALG5500-1G SIALG5500-250MG SIALG5500-25G SIALG5500-500MG
L(-)-Glucose, Sigma-Aldrich®
L(-)-Glucose
Vzorec: C₆H₁₂O₆
Bod varu: 376 °C (1013 hPa)
Bod tání: 146…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00148913
CAS číslo: 921-60-8
EINECS: 213-068-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR