L(-)-Galactose ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(2S,3R,4R,5S)-2,3,4,5,6-Pentahydroxyhexanal, (2S,3R,4R,5S)-Pentahydroxyhexanal, L-Galactose
B21448.MC
B21448.ME
B21448.MCEA
3510
CZK
B21448.MC
B21448.ME
L(-)-Galactose ≥98%
L(-)-Galactose
Purity: 98%
CAS No.: 15572-79-9
Molecular Formula: C6H12O6
Formula Weight: 180.16
Melting Point: 163-165°
Beilstein Reference: 1724622
Vzorec:
C₆H₁₂O₆ MW: 180.16 Bod tání: 163…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063833 CAS číslo: 15572-79-9 EINECS: 239-630-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...