Tisk…

L(+)-Arabinóza ≥99%

Dodavatel: Thermo Fisher Scientific

A11921.18 A11921.30
A11921.18EA 2860 CZK
A11921.18 A11921.30
L(+)-Arabinóza ≥99%
L(+)-Arabinóza

Purity: 99% CAS No.: 87-72-9 Molecular Formula: C5H10O5 Formula Weight: 150.13 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 155-160° Beilstein Reference: 1723085 Merck Reference: 14,761 Optical Rotation: +104° (c=10 in water, 24h) MDL No.: MFCD00067709

Vzorec: C₅H₁₀O₅
Bod varu: 331 °C (1013 hPa)
Bod tání: 155…160 °C
Hustota: 1,585 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS číslo: 5328-37-0
EINECS: 201-767-6
Merck Index: 14,761
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR