Tisk…

D(+)-Glukóza, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

M-Clarity™ quality level = MQ200

G6152-100G G6152-1KG G6152-500G
SIALG6152-100GEA 3390 CZK
SIALG6152-100G SIALG6152-1KG SIALG6152-500G
D(+)-Glukóza, Sigma-Aldrich®
D(+)-Glukóza
Vzorec: C₆H₁₂O₆
MW: 180.16
Bod tání: 146
Hustota: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS číslo: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR