Tisk…

β-D-Glucose ≥80%, crystalline powder

Dodavatel: MP Biomedicals

Synonyms: beta-D-Glucose

0210095391 0210095310 0210095325 0210095380
ICNA0210095391EA 0 CZK
ICNA0210095391 ICNA0210095310 ICNA0210095325 ICNA0210095380
β-D-Glucose ≥80%, crystalline powder
β-D-Glucose

β-D-Glucose is a main source of energy for living organisms. It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen. It may contain up to 10% of α-isomer.


Soluble in water (1/20-clear solution).

Vzorec: C₆H₁₂O₆
MDL Number: MFCD00063989
CAS číslo: 492-61-5
EINECS: 207-756-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR