D-(+)-Sucrose octaacetate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W303801-10KG-K
W303801-1KG-K
W303801-25KG-K
SIALW303801-10KG-KEA
21840
CZK
SIALW303801-10KG-K
SIALW303801-1KG-K
SIALW303801-25KG-K
D-(+)-Sucrose octaacetate, Sigma-Aldrich®
D-(+)-Sucrose octaacetate
Vzorec:
C₂₈H₃₈O₁₉ Bod tání: 80…83 °C Hustota: 1,257 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006623 CAS číslo: 126-14-7 EINECS: 204-772-1 Merck Index: 12,09052 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...