D-(+)-Sucrose octaacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
D-(+)-Saccharose octaacetate, Sucrose octaacetate, D(+)-Sucroseoctaacetat, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(((2S,3S,4R,5R)-3,4-diacetoxy-2,5-bis(acetoxymethyl)tetrahydrofuran-2-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
M-Clarity™ quality level = MQ200
252603-1KG
252603-250G
SIAL252603-1KGEA
4810
CZK
SIAL252603-1KG
SIAL252603-250G
D-(+)-Sucrose octaacetate, Sigma-Aldrich®
D-(+)-Sucrose octaacetate
Vzorec:
C₂₈H₃₈O₁₉ MW: 678,6 g/mol Bod tání: 80…83 °C Hustota: 1,257 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006623 CAS číslo: 126-14-7 EINECS: 204-772-1 Merck Index: 12,09052 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...