D(+)-Rafinóza pentahydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate
M-Clarity™ quality level = MQ200
R7630-100G
R7630-500G
SIALR7630-100GEA
22180
CZK
SIALR7630-100G
SIALR7630-500G
D(+)-Rafinóza pentahydrát, Sigma-Aldrich®
D(+)-Rafinóza pentahydrát
Vzorec:
C₁₈H₃₂O₁₆·5H₂O Bod tání: 80 °C Hustota: 1,516 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071590 CAS číslo: 17629-30-0 EINECS: 208-146-9 Merck Index: 12,08279 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...