D(+)-Trehalóza dihydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
M-Clarity™ quality level = MQ200
T9449-100G
T9449-25G
T9449-500G
SIALT9449-100GEA
14560
CZK
SIALT9449-100G
SIALT9449-25G
SIALT9449-500G
D(+)-Trehalóza dihydrát, Sigma-Aldrich®
D(+)-Trehalóza dihydrát
Vzorec:
C₁₂H₂₂O₁₁·2H₂O Bod tání: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00071594 CAS číslo: 6138-23-4 EINECS: 202-739-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...