Tisk…

DL-Arabinose, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

A9524-10G A9524-50G
SIALA9524-10GEA 6000 CZK
SIALA9524-10G SIALA9524-50G
DL-Arabinose, Sigma-Aldrich®
DL-Arabinose
Vzorec: C₅H₁₀O₅
MW: 150,13 g/mol
Bod varu: 331 °C (1013 hPa)
Bod tání: 159…160 °C
Hustota: 1,609 g/cm³ (17 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135867
CAS číslo: 147-81-9
EINECS: 205-699-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR