Tisk…

DL-Arabinose ≥98%

Dodavatel: Thermo Fisher Scientific

L09895.09 L09895.18
L09895.09EA 2170 CZK
L09895.09 L09895.18
DL-Arabinose ≥98%
DL-Arabinose

Purity: 98+% CAS No.: 147-81-9 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 158-160° Beilstein Reference: 1723086 Merck Reference: 14,761 MDL No.: MFCD00135867

Vzorec: C₅H₁₀O₅
Bod varu: 331 °C (1013 hPa)
Bod tání: 158…160 °C
Hustota: 1,609 g/cm³ (17 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135867
CAS číslo: 147-81-9
EINECS: 205-699-8
Merck Index: 14,761

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR