Fullerene C70 99+%, powder
Dodavatel: Thermo Scientific
|
Varování
|
42600.03
42600.MD
42600.03EA
45950
CZK
42600.03
42600.MD
Fullerene C70 99+%, powder
Fullerene C70
Purity: 99+%
CAS No.: 115383-22-7
Physical Form: Powder
Density: 1.7
Appearance: Grey/black
Boiling Point: 500-600° subl.
UN No.: 1325
MDL No.: MFCD00282904
Vzorec:
C₇₀ Bod varu: 500…600 °C (subl.) Bod tání: > 280 °C Hustota: 1,7 Storage Temperature: Ambient |
MDL Number:
MFCD00282904 CAS číslo: 115383-22-7 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...