Fullerene C60, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
483036-1G
483036-5G
SIAL483036-1GEA
7560
CZK
SIAL483036-1G
SIAL483036-5G
Fullerene C60, Sigma-Aldrich®
Fullerene C60
Vzorec:
C₆₀ Bod tání: > 280 °C Hustota: ∼1,65 g/cm³ (20 °C) |
MDL Number:
MFCD00151408 CAS číslo: 99685-96-8 UN: 1325 ADR: 4.1,III Merck Index: 13,01447 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...