Tisk…

Fullerene C60, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen

M-Clarity™ quality level = MQ100

379646-1G 379646-5G
SIAL379646-1GEA 10620 CZK
SIAL379646-1G SIAL379646-5G
Fullerene C60, Sigma-Aldrich®
Fullerene C60
Vzorec: C₆₀
Bod tání: > 280 °C
Hustota: ∼1,65 g/cm³ (20 °C)
MDL Number: MFCD00151408
CAS číslo: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR