Tisk…

Fullerene C60 ≥99.9%, powder

Dodavatel: Thermo Scientific
Varování

42007.03 42007.MD
42007.03EA 15960 CZK
42007.03 42007.MD
Fullerene C60 ≥99.9%, powder
Fullerene C60

Purity: 99.9+% CAS No.: 99685-96-8 Density: 1.7 Appearance: Black Boiling Point: 500-600° subl. UN No.: 1325 MDL No.: MFCD00282904

Vzorec: C₆₀
Bod varu: 500…600 °C (subl.)
Bod tání: > 280 °C
Hustota: 1,7
Storage Temperature: Ambient
MDL Number: MFCD00151408
CAS číslo: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR