Deuteriumoxid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Water-D2, Heavy water
M-Clarity™ quality level = MQ100
450510-100ML
450510-25ML
450510-10X0.75
SIAL450510-100MLEA
7910
CZK
SIAL450510-100ML
SIAL450510-25ML
SIAL450510-10X0.75
Deuteriumoxid, Sigma-Aldrich®
Deuteriumoxid
Vzorec:
D₂O Bod varu: 101,3 °C (1013 hPa) Hustota: 1,1 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044636 CAS číslo: 7789-20-0 EINECS: 232-148-9 Merck Index: 12,02984 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...