Tisk…

Cyclohexane-[D₁₂], Sigma-Aldrich®

Dodavatel: Merck

151866-10G 151866-1G 151866-5G
SIAL151866-10GEA 8040 CZK
SIAL151866-10G SIAL151866-1G SIAL151866-5G
Cyclohexane-[D₁₂], Sigma-Aldrich®
Cyclohexane-[D₁₂]
Vzorec: C₆D₁₂
MDL Number: MFCD00044212
CAS číslo: 1735-17-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR