Tisk…

Benzene-[D₁], Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

175722-1G 175722-5G
SIAL175722-1GEA 4300 CZK
SIAL175722-1G SIAL175722-5G
Benzene-[D₁], Sigma-Aldrich®
Benzene-[D₁]
Vzorec: C₆H₅D
CAS číslo: 1120-89-4
EINECS: 214-321-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR