Phenol-[D₆], Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Monohydroxybenzene-D6, Hydroxybenzene-D6, Phenylalcohol-D6, Phenol-[²H₆], Carbolic acid-D6
M-Clarity™ quality level = MQ200
176060-1G
176060-5G
SIAL176060-1GEA
5270
CZK
SIAL176060-1G
SIAL176060-5G
Phenol-[D₆], Sigma-Aldrich®
Phenol-[D₆]
Vzorec:
C₆D₅OD Bod varu: 180 °C (1013 hPa) Hustota: 1,06 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS číslo:
13127-88-3 EINECS: 236-063-8 UN: 1671 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...