Tisk…

Aniline-[D₇], Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Phenylamine-D7, Aniline-[²H₇], Aminobenzene-D7

M-Clarity™ quality level = MQ200

175706-1G 175706-5G
SIAL175706-1GEA 4230 CZK
SIAL175706-1G SIAL175706-5G
Aniline-[D₇], Sigma-Aldrich®
Aniline-[D₇]
Vzorec: C₆D₅ND₂
CAS číslo: 14545-23-4
EINECS: 238-580-4
UN: 1547
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR