Tisk…

1,4-Dichlorobenzene-[D₄], Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,4-Dichlorobenzene-[²H₄]

M-Clarity™ quality level = MQ200

329339-1G 329339-5G
SIAL329339-1GEA 4210 CZK
SIAL329339-1G SIAL329339-5G
1,4-Dichlorobenzene-[D₄], Sigma-Aldrich®
1,4-Dichlorobenzene-[D₄]
Vzorec: C₆D₄Cl₂
MW: 150,97 g/mol
CAS číslo: 3855-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR