Tisk…
Anthracene-[D₁₀], Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
176591-1G 176591-5G
SIAL176591-1GEA 6140 CZK
SIAL176591-1G SIAL176591-5G
Anthracene-[D₁₀], Sigma-Aldrich®
Anthracene-[D₁₀]
Vzorec: C₁₄D₁₀
Bod varu: 340 °C (1013 hPa)
Storage Temperature: Refrigerator
CAS číslo: 1719-06-8
EINECS: 217-004-5