Dimethyl sulphate-[D₆], Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
164526-10G
164526-1G
SIAL164526-10GEA
13810
CZK
SIAL164526-10G
SIAL164526-1G
Dimethyl sulphate-[D₆], Sigma-Aldrich®
Dimethyl sulphate-[D₆]
Vzorec:
D₃CS(O)₂CD₃ Bod varu: 188 °C (1013 hPa) Bod tání: –32 °C Hustota: 1,39 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS číslo:
15199-43-6 EINECS: 239-257-0 UN: 1595 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...