Tisk…

Aceton-D6, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6

M-Clarity™ quality level = MQ100

454133-25G 454133-50G
SIAL454133-25GEA 3860 CZK
SIAL454133-25G SIAL454133-50G
Aceton-D6, Sigma-Aldrich®
Aceton-D6
Vzorec: D₃CC(O)CD₃
Bod varu: 55,5 °C (1013 hPa)
Hustota: 0,79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS číslo: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR