Tisk…

Petrolether, 60…80 °C, AnalaR NORMAPUR® zA

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

23840.292EA 1730 CZK
23840.292 23840.326 23840.460
Petrolether, 60…80 °C, AnalaR NORMAPUR® zA
Ropný líh
Bod varu: 60…80 °C (1013 hPa)
Hustota: 0.67 g/cm³ (20 °C)
Bod vzplanutí: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS číslo: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Objednat

Výsledky specifikačního testu

Acidity Max. 0.0002 meq/g
Bromine value Max. 3
Colouration Max. 10 APHA
Substances coloured by H2SO4 Max. 150 APHA
Boiling range (60-80°C) Min. 90 %
Evaporation residue Max. 10 ppm
Sulphur compounds (as CS2) Max. 3 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.15 ppm
Ba (Barium) Max. 0.08 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.3 ppm
Mg (Magnesium) Max. 0.08 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.75 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.08 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.03 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR