Tisk…
Petrolether, 40…60 °C, CHROMASOLV™ Pest_Residual, hydrogen treated naphtha_low boiling pt, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Nebezpečí
Synonyms: Gasoline, Petroleum ether, Petrol, Benzine
34491-45L 34491-7L 34491-1L 34491-2.5L 34491-4X2.5L
34491-45LEA 83530 CZK
34491-45L 34491-7L 34491-1L 34491-2.5L 34491-4X2.5L
Petrolether, 40…60 °C, CHROMASOLV™ Pest_Residual, hydrogen treated naphtha_low boiling pt, Riedel-de Haën™
Ropný líh
  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Bod varu: 40…60 °C (1.013 hPa)
Hustota: 0.65 g/cm³ (20 °C)
Bod vzplanutí: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS číslo: 64742-49-0
EINECS: 309-852-0
UN: 3295
ADR: 3 II
Objednat
Výsledky specifikačního testu
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l