Petrolether, 40…60 °C, extra pure
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Gasoline, Petroleum ether, Petrol, Benzine
124860025
124860050
124860250
ACRO124860025EA
3250
CZK
ACRO124860025
ACRO124860050
ACRO124860250
Petrolether, 40…60 °C, extra pure
Ropný líh
Bod varu:
40…60 °C (760 mmHg) Hustota: 0,65 g/cm³ Bod vzplanutí: –55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS číslo: 64742-49-0 EINECS: 265-151-9 UN: 3295 ADR: 3,II |
Výsledky specifikačního testu
Appearance | Clear liquid |
Water | ≤0.03 % (K.F.) |
Boiling point | 40 to 60 °C |
Density | (20 °C) 0.64 to 0.66 g/ml |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...