Tisk…
parafín, Solidification point: 54-56°C, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Wax Paraffin, Paraffin wax
M-Clarity™ quality level = MQ200
327204-12KG 327204-1KG
SIAL327204-12KGEA 21970 CZK
SIAL327204-12KG SIAL327204-1KG
parafín, Solidification point: 54-56°C, Sigma-Aldrich®
Parafín
CAS číslo: 8002-74-2
EINECS: 232-315-6