4-Methylcyclohexanol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
153095-1L
153095-250ML
SIAL153095-1LEA
5970
CZK
SIAL153095-1L
SIAL153095-250ML
4-Methylcyclohexanol, Sigma-Aldrich®
4-Methylcyclohexanol
Vzorec:
C₇H₁₄O Bod varu: 171 °C (1013 hPa) Bod tání: –50 °C Hustota: 0,9328 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001449 CAS číslo: 589-91-3 EINECS: 209-664-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...