Tisk…

Ethyl propenyl ether, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 1-Propenyl ether

364045-10G 364045-25G
SIAL364045-10GEA 3740 CZK
SIAL364045-10G SIAL364045-25G
Ethyl propenyl ether, Sigma-Aldrich®
Ethyl propenyl ether
Vzorec: C₅H₁₀O
MW: 86,13 g/mol
Bod varu: 55…65 °C (1013 hPa)
Hustota: 0,7951 g/cm³ (0 °C)
MDL Number: MFCD00015184
CAS číslo: 928-55-2
EINECS: 213-176-0
UN: 3271
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR