Tisk…

2-Bromo-2-butene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

215562-100G 215562-25G
SIAL215562-100GEA 7800 CZK
SIAL215562-100G SIAL215562-25G
2-Bromo-2-butene, Sigma-Aldrich®
2-Bromo-2-butene
Vzorec: C₄H₇Br
MW: 135 g/mol
Bod varu: 82 °C (1013 hPa)
Hustota: 1,32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000141
CAS číslo: 13294-71-8
EINECS: 236-316-2
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR