Tisk…

Rhodium, foil, Thickness: 0.025mm (0.001 in)

Dodavatel: Thermo Fisher Scientific

11524.FF 11524.FI
11524.FFEA 44370 CZK
11524.FF 11524.FI
Rhodium, foil, Thickness: 0.025mm (0.001 in)
Rhodium
Vzorec: Rh
Bod varu: 3400…3700 °C (1013 hPa)
Bod tání: 1960 °C
Hustota: 12,41 g/cm³ (20 °C)
MDL Number: MFCD00011201
CAS číslo: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR