Platina, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Platinium
MQ100
267244-1.4G
267244-350MG
SIAL267244-1.4GEA
21600
CZK
SIAL267244-1.4G
SIAL267244-350MG
Platina, Sigma-Aldrich®
Platina
Vzorec:
Pt MW: 195.08 Bod varu: 3827 Bod tání: 1769 Hustota: 21.447 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS číslo: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...