Tisk…

Platina ≥99.99% (metals basis), wire, stress relieved, Ø 0.0508 mm (0.002 in)

Dodavatel: Thermo Fisher Scientific

Synonyms: Platinium

43772.G1 43772.G5
43772.G1EA 3320 CZK
43772.G1 43772.G5
Platina ≥99.99% (metals basis), wire, stress relieved, Ø 0.0508 mm (0.002 in)
Platina

Purity: 99.99% CAS No.: 7440-06-4 Physical Form: ≈0.043g/m, Temper: soft Odour: Odourless MDL No.: MFCD00011179

Vzorec: Pt
Bod varu: 3827 °C (1013 hPa)
Bod tání: 1769 °C
Hustota: 21,447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR