Tisk…

Platina ≥99.9% (metals basis), powder -325 mesh

Dodavatel: Thermo Fisher Scientific

Synonyms: Platinium

38374.03 38374.06 38374.77
38374.03EA 9410 CZK
38374.03 38374.06 38374.77
Platina ≥99.9% (metals basis), powder -325 mesh
Platina

Purity: 99.9+% CAS No.: 7440-06-4 Odour: Odourless MDL No.: MFCD00011179

Vzorec: Pt
Bod varu: 4530 °C (1013 hPa)
Bod tání: 1773 °C
Hustota: 21,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR