Tisk…

Platina ≥99.997% (metals basis), wire, Premion®, Ø 1.0 mm (0.04 in)

Dodavatel: Thermo Fisher Scientific

10956.BP 10956.BS 10956.BW
10956.BPEA 5740 CZK
10956.BP 10956.BS 10956.BW
Platina ≥99.997% (metals basis), wire, Premion®, Ø 1.0 mm (0.04 in)
Platina

Purity: 99.997% CAS No.: 7440-06-4 Physical Form: ≈16.8g/m Odour: Odourless MDL No.: MFCD00011179

Vzorec: Pt
Bod varu: 3827 °C (1013 hPa)
Bod tání: 1769 °C
Hustota: 21,447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR