Tisk…

Platina ≥99.99% (metals basis), powder, Premion® -22 mesh

Dodavatel: Thermo Fisher Scientific

10672.01 10672.03 10672.06 10672.14
10672.01EA 3850 CZK
10672.01 10672.03 10672.06 10672.14
Platina ≥99.99% (metals basis), powder, Premion® -22 mesh
Platina

Purity: 99.99% CAS No.: 7440-06-4 Appearance: Silver-white Odour: Odourless MDL No.: MFCD00011179

Vzorec: Pt
Bod varu: 3827 °C (1013 hPa)
Bod tání: 1769 °C
Hustota: 21,447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR