Tisk…
Platina 99,9 Metals basis, wire, hard, Ø 0.127 mm (0.005 in)
Dodavatel: Thermo Fisher Scientific
Synonyms: Platinium
00263.G1 00263.G5
00263.G1EA 3950 CZK
00263.G1 00263.G5
Platina 99,9 Metals basis, wire, hard, Ø 0.127 mm (0.005 in)
Platina
Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈0.27g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011179
Vzorec: Pt
MW: 195.08
Bod varu: 3827
Bod tání: 1769
Hustota: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689