Tisk…

Paladium ≥99.95% (metals basis), wire, hard, Ø 0.203 mm

Dodavatel: Thermo Fisher Scientific

45072.G1 45072.G6
45072.G1EA 1800 CZK
45072.G1 45072.G6
Paladium ≥99.95% (metals basis), wire, hard, Ø 0.203 mm
Palladium

Purity: 99.95% CAS No.: 7440-05-3 Odour: Odourless MDL No.: MFCD00011167

Vzorec: Pd
Bod varu: 3167 °C (1013 hPa)
Bod tání: 1555 °C
Hustota: 12,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR