Paladium ≥99.99% (metals basis), wire, Premion®, Ø 0.25 mm
Dodavatel: Thermo Fisher Scientific
10962.G1
10962.G5
10962.G9
10962.G1EA
6830
CZK
10962.G1
10962.G5
10962.G9
Paladium ≥99.99% (metals basis), wire, Premion®, Ø 0.25 mm
Palladium
Purity: 99.99%
CAS No.: 7440-05-3
Physical Form: ≈0.6g/m
Odour: Odourless
MDL No.: MFCD00011167
Vzorec:
Pd MW: 106.420 g/mol Hustota: 12.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS číslo: 7440-05-3 EINECS: 231-115-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...