Nikl, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
267007-1.4G
267007-5.6G
SIAL267007-1.4GEA
7660
CZK
SIAL267007-1.4G
SIAL267007-5.6G
Nikl, Sigma-Aldrich®
Nikl
Vzorec:
Ni MW: 58.69 Bod varu: 3007 Bod tání: 1453 Hustota: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS číslo: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1 III Merck Index: 12,06582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...