Tisk…
Nikl ≥99.9945% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Dodavatel: Thermo Fisher Scientific
Nebezpečí
12042.FF 12042.FI
12042.FFEA 6040 CZK
12042.FF 12042.FI
Nikl ≥99.9945% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Nikl
Purity: 99.9945% CAS No.: 7440-02-0 Physical Form: ≈11.1g/25x25mm Odour: Odourless MDL No.: MFCD00011137
Vzorec: Ni
MW: 58.69
Bod varu: 3007
Bod tání: 1453
Hustota: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS číslo: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582