Tisk…

Nikl ≥99.9% (metals basis), pieces

Dodavatel: Thermo Scientific
Nebezpečí

00434.22 00434.30
00434.22EA 2220 CZK
00434.22 00434.30
Nikl ≥99.9% (metals basis), pieces
Nikl

Purity: 99.9+% CAS No.: 7440-02-0 Odour: Odourless MDL No.: MFCD00011137

Vzorec: Ni
MW: 58,69 g/mol
Bod varu: 3007 °C (1013 hPa)
Bod tání: 1453 °C
Hustota: 8,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS číslo: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR