Tisk…

Iridium ≥99.95% (metals basis), powder -60 mesh

Dodavatel: Thermo Scientific

12070.02 12070.04 12070.09
12070.02EA 14010 CZK
12070.02 12070.04 12070.09
Iridium ≥99.95% (metals basis), powder -60 mesh
Iridium

Purity: 99.95% CAS No.: 7439-88-5 Appearance: Grey MDL No.: MFCD00011062

Vzorec: Ir
MW: 192,22 g/mol
Bod varu: 4500 °C (1013 hPa)
Bod tání: 2443 °C
Hustota: 22,65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS číslo: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III
Merck Index: 12,05098

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR