Železo ≥99.97% (metals basis), pieces, irregularly shaped ≤ 25 mm (1.0 in)
Dodavatel: Thermo Fisher Scientific
10213.30
10213.A1
10213.A7
10213.PG_25KG
10213.30EA
1670
CZK
10213.30
10213.A1
10213.A7
10213.PG_25KG
Železo ≥99.97% (metals basis), pieces, irregularly shaped ≤ 25 mm (1.0 in)
Železo
Purity: 99.97+%
CAS No.: 7439-89-6
Physical Form: 90% 2.5-25mm (0.1-1.0in)
Appearance: Gray
MDL No.: MFCD00010999
Vzorec:
Fe Bod varu: 3000 °C (1013 hPa) Bod tání: 1535 °C Hustota: 7,86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS číslo: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...