Kobalt, powder, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
266647-250G
266647-50G
SIAL266647-250GEA
10720
CZK
SIAL266647-250G
SIAL266647-50G
Kobalt, powder, Sigma-Aldrich®
Kobalt
Vzorec:
Co |
MDL Number:
MFCD00010935 CAS číslo: 7440-48-4 EINECS: 231-158-0 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...