Tisk…

Kobalt, powder, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

266639-100G 266639-500G
SIAL266639-100GEA 4490 CZK
SIAL266639-100G SIAL266639-500G
Kobalt, powder, Sigma-Aldrich®
Kobalt
Vzorec: Co
MDL Number: MFCD00010935
CAS číslo: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR